accessibility menu, dialog, popup

UserName

CAS RN 10022-13-6

IUPAC Name: [3,4,6-tris(acetyloxy)-5-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)oxan-2-yl]methyl acetate
Synonyms: [3,4,6-tris(acetyloxy)-5-(1,3-dioxoisoindol-2-yl)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 477.42
Molecular Formula: C22H23NO11
InChi Key: DUXJAHFLYZUOPT-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(C)=O)C(C(OC(C)=O)C1OC(C)=O)N1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results