missing translation for 'onlineSavingsMsg'
Learn More

1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranose 97.0+%, TCI America™

Catalog No. p-7137078
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20475G 5 g
T204725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20475G Supplier TCI America Supplier No. T20475G

Chemical Identifiers

CAS 10022-13-6
Molecular Formula C22H23NO11
Molecular Weight (g/mol) 477.422
InChI Key DUXJAHFLYZUOPT-ACMVSEJYSA-N
PubChem CID 2728904
IUPAC Name [(2R,3S,4R,5R,6S)-3,4,6-triacetyloxy-5-(1,3-dioxoisoindol-2-yl)oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)N2C(=O)C3=CC=CC=C3C2=O)OC(=O)C)OC(=O)C

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 5 g
Formula Weight 477.42
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranose
TSCA No
Recommended Storage Freezer