missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine, Thermo Scientific™

Chemical Identifiers

CAS 859833-21-9
Molecular Formula C18H28BNO2
Molecular Weight (g/mol) 301.24
MDL Number MFCD08435871
InChI Key KEKNXVTYDJCRMT-UHFFFAOYSA-N
Synonym 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 16414184
IUPAC Name 1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine
SMILES CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC45239CB
View Documents Promotion Details
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45239DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45239DE
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

859833-21-9
301.24
KEKNXVTYDJCRMT-UHFFFAOYSA-N
16414184
CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1
C18H28BNO2
MFCD08435871
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine

Specifications

859833-21-9
100.0
Amber Glass Bottle
MFCD08435871
KEKNXVTYDJCRMT-UHFFFAOYSA-N
1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine
16414184
97%
96.5
97%
C18H28BNO2
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1
301.24
301.24
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents