Learn More
1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC45239DE
| Quantity | 5g |
|---|
Chemical Identifiers
| 859833-21-9 | |
| 301.24 | |
| KEKNXVTYDJCRMT-UHFFFAOYSA-N | |
| 16414184 | |
| CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1 |
| C18H28BNO2 | |
| MFCD08435871 | |
| 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine |
Specifications
| 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine | |
| 96.5 | |
| 97% | |
| 5g | |
| 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1 | |
| 301.24 | |
| 301.24 |
| 859833-21-9 | |
| 100.0 | |
| C18H28BNO2 | |
| MFCD08435871 | |
| KEKNXVTYDJCRMT-UHFFFAOYSA-N | |
| 1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine | |
| 16414184 | |
| 97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.