missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC45239DA

Quantity 1g

 View more versions of this product

Catalog No. CC45239DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

859833-21-9
301.24
KEKNXVTYDJCRMT-UHFFFAOYSA-N
16414184
CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1
C18H28BNO2
MFCD08435871
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine

Specifications

1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine
96.5
97%
C18H28BNO2
MFCD08435871
KEKNXVTYDJCRMT-UHFFFAOYSA-N
1-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]piperidine
16414184
97%
859833-21-9
100.0
Amber Glass Bottle
1g
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, piperidine,1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, 3-piperidine methyl phenylboronic acid pinacol ester, amtb771, 3-piperidin-1-ylmethyl phenylboronic acid, pinacol ester, 2-3-piperidinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC(CN2CCCCC2)=CC=C1
301.24
301.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions