missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,2-Dioxathiolane 2,2-Dioxide 98.0+%, TCI America™
Supplier: TCI America D283025G
Specifications
| 1,3,2-Dioxathiolane 2,2-Dioxide | |
| 98°C | |
| C2H4O4S | |
| 25 g | |
| ZPFAVCIQZKRBGF-UHFFFAOYSA-N | |
| 1,3,2-dioxathiolane 2,2-dioxide | |
| 14075 | |
| ≥98.0% (GC) |
| 1072-53-3 | |
| White-Yellow | |
| MFCD00221769 | |
| ethylenesulfate, ethylene sulfate, ethylene glycol, cyclic sulfate, glycol sulfate, 1,2-ethylene sulfate, 1,3,2-dioxathiolane, 2,2-dioxide, sulfuric acid, cyclic ethylene ester, 1,3,2-dioxathiolane-2,2-dioxide, 1,3,2??-dioxathiolane-2,2-dione, ethosulfate | |
| C1COS(=O)(=O)O1 | |
| 124.11 | |
| 124.11 | |
| Crystalline Powder |
Chemical Identifiers
| 1072-53-3 | |
| 124.11 | |
| ZPFAVCIQZKRBGF-UHFFFAOYSA-N | |
| 14075 | |
| C1COS(=O)(=O)O1 |
| C2H4O4S | |
| MFCD00221769 | |
| ethylenesulfate, ethylene sulfate, ethylene glycol, cyclic sulfate, glycol sulfate, 1,2-ethylene sulfate, 1,3,2-dioxathiolane, 2,2-dioxide, sulfuric acid, cyclic ethylene ester, 1,3,2-dioxathiolane-2,2-dioxide, 1,3,2??-dioxathiolane-2,2-dione, ethosulfate | |
| 1,3,2-dioxathiolane 2,2-dioxide |
Safety and Handling
RTECSNumber : JH4800000
TSCA : No