accessibility menu, dialog, popup

UserName

CAS RN 1072-53-3

IUPAC Name: 1,3,2λ⁶-dioxathiolane-2,2-dione
Synonyms: 1,3,2λ⁶-dioxathiolane-2,2-dione
Molecular Weight (g/mol): 124.11
Molecular Formula: C2H4O4S
InChi Key: ZPFAVCIQZKRBGF-UHFFFAOYSA-N
SMILES: O=S1(=O)OCCO1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)