missing translation for 'onlineSavingsMsg'
Learn More

1,3,2-Dioxathiolane 2,2-Dioxide 98.0+%, TCI America™

Catalog No. p-7127524
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28305G 5 g
D283025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28305G Supplier TCI America Supplier No. D28305G

Chemical Identifiers

CAS 1072-53-3
Molecular Formula C2H4O4S
Molecular Weight (g/mol) 124.11
MDL Number MFCD00221769
InChI Key ZPFAVCIQZKRBGF-UHFFFAOYSA-N
Synonym ethylenesulfate, ethylene sulfate, ethylene glycol, cyclic sulfate, glycol sulfate, 1,2-ethylene sulfate, 1,3,2-dioxathiolane, 2,2-dioxide, sulfuric acid, cyclic ethylene ester, 1,3,2-dioxathiolane-2,2-dioxide, 1,3,2??-dioxathiolane-2,2-dione, ethosulfate
PubChem CID 14075
IUPAC Name 1,3,2-dioxathiolane 2,2-dioxide
SMILES C1COS(=O)(=O)O1

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 5 g
Formula Weight 124.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,2-Dioxathiolane 2,2-Dioxide
RTECSNumber JH4800000
TSCA No