missing translation for 'onlineSavingsMsg'
Learn More

1,2-Naphthoquinone 95.0+%, TCI America™

Catalog No. N00411G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N00411G 1 g
N00415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. N00411G missing translation for 'mfr' TCI America missing translation for 'supplierNo' N00411G

Chemical Identifiers

CAS 524-42-5
Molecular Formula C10H6O2
Molecular Weight (g/mol) 158.16
MDL Number MFCD00001698
InChI Key KETQAJRQOHHATG-UHFFFAOYSA-N
Synonym 1,2-naphthoquinone, 1,2-naphthalenedione, o-naphthoquinone, beta-naphthoquinone, 1,2-naphthaquinone, 1,2-naftochinon, .beta.-naphthoquinone, 1,2-naftochinon czech, ccris 1558, 1,2-dihydronaphthalene-1,2-dione
PubChem CID 10667
ChEBI CHEBI:34055
IUPAC Name 1,2-dihydronaphthalene-1,2-dione
SMILES O=C1C=CC2=CC=CC=C2C1=O

Specifications

Melting Point 133°C
Quantity 1 g
Formula Weight 158.16
Percent Purity ≥95.0% (HPLC,W)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Naphthoquinone
EINECSNumber (4)-0372
RTECSNumber QL7000000
TSCA Yes
Recommended Storage Freezer