accessibility menu, dialog, popup

UserName

OO

CAS RN 524-42-5

IUPAC Name: 1,2-dihydronaphthalene-1,2-dione
Synonyms: 1,2-naphthoquinone 1,2-naphthaquinone 1,2-naftochinon β-naphthoquinone
Molecular Weight (g/mol): 158.16
Molecular Formula: C10H6O2
InChi Key: KETQAJRQOHHATG-UHFFFAOYSA-N
SMILES: O=C1C=CC2=CC=CC=C2C1=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)