missing translation for 'onlineSavingsMsg'
Learn More

1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%

Catalog No. AC415400100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415400100 10 g Glass bottle
AC415400250 25 g Glass bottle
AC415401000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC415400100 Supplier Thermo Scientific Chemicals Supplier No. 415400100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 521-24-4 | C10H5NaO5S | 260.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 521-24-4
Molecular Formula C10H5NaO5S
Molecular Weight (g/mol) 260.19
MDL Number MFCD00001700
InChI Key UBLXEEBHYISRFM-UHFFFAOYSA-M
Synonym 1,2-naphthoquinone-4-sulfonic acid sodium salt, sodium 1,2-naphthoquinone-4-sulfonate, sodium beta-naphthoquinone-4-sulfonate, sodium 3,4-dioxonaphthalene-1-sulfonate, sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, unii-t7185hp5sh, sodium 3,4-dioxonaphthalene-1-sulphonate, beta-naphthoquinone-4-sulfonate sodium salt, 1-naphthalenesulfonic acid, 3,4-dihydro-3,4-dioxo-, sodium salt, chembl61245
PubChem CID 516996
IUPAC Name sodium;3,4-dioxonaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=O)S(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 289.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Quantity 10 g
Beilstein 02, 330
Merck Index 15, 8777
Solubility Information Solubility in water: 33.3g/L (20°C)
Formula Weight 260.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 2-Naphthoquinone-4-sulfonic acid sodium salt, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-308-9
RTECSNumber QK1490000
TSCA TSCA

RUO – Research Use Only