Learn More
1-(2-Fluorophenyl)-2-propylamine, 97%
CAS: 1716-60-5 | C9H12FN | 153.20 g/mol
Supplier: Thermo Scientific Chemicals H5099903
Description
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as intermediate for pharmaceutical.
Solubility
Slightly Soluble (2.0 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 1-(2-Fluorophenyl)-2-propylamine | |
| C9H12FN | |
| 1 g | |
| Slightly Soluble (2.0g/L) (25°C). | |
| CC(N)CC1=CC=CC=C1F | |
| 153.20 | |
| 153.2 |
| 1716-60-5 | |
| MFCD01708100 | |
| 2-fluoroamphetamine, 1-2-fluorophenyl propan-2-amine, o-fluoro-alpha-methylphenethylamine, phenethylamine, o-fluoro-alpha-methyl, 1-2-fluorophenyl-2-propylamine, 2-fa, o-fluoro-a-methylphenethylamine, benzeneethanamine,2-fluoro-a-methyl, 1-2-fluorophenyl propan-2-aminehydrochloride | |
| GDSXNLDTQFFIEU-UHFFFAOYNA-N | |
| 1-(2-fluorophenyl)propan-2-amine | |
| 121531 | |
| 97% |
Chemical Identifiers
| 1716-60-5 | |
| 153.20 | |
| GDSXNLDTQFFIEU-UHFFFAOYNA-N | |
| 121531 | |
| CC(N)CC1=CC=CC=C1F |
| C9H12FN | |
| MFCD01708100 | |
| 2-fluoroamphetamine, 1-2-fluorophenyl propan-2-amine, o-fluoro-alpha-methylphenethylamine, phenethylamine, o-fluoro-alpha-methyl, 1-2-fluorophenyl-2-propylamine, 2-fa, o-fluoro-a-methylphenethylamine, benzeneethanamine,2-fluoro-a-methyl, 1-2-fluorophenyl propan-2-aminehydrochloride | |
| 1-(2-fluorophenyl)propan-2-amine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : SH7030000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only