CAS RN 1716-60-5
IUPAC Name:
1-(2-fluorophenyl)propan-2-amine
Synonyms:
2-fluoroamphetamine
Molecular Weight (g/mol):
153.2
Molecular Formula:
C9H12FN
InChi Key:
GDSXNLDTQFFIEU-UHFFFAOYNA-N
SMILES:
CC(N)CC1=CC=CC=C1F
1
–
1
of
1
results
1
Promotions Available
1-(2-Fluorophenyl)-2-propylamine, 97%
1-(2-Fluorophenyl)-2-propylamine, 97%
CAS: 1716-60-5 Molecular Formula: C9H12FN Molecular Weight (g/mol): 153.20 MDL Number: MFCD01708100 InChI Key: GDSXNLDTQFFIEU-UHFFFAOYNA-N Synonym: 2-fluoroamphetamine,1-2-fluorophenyl propan-2-amine,o-fluoro-alpha-methylphenethylamine,phenethylamine, o-fluoro-alpha-methyl,1-2-fluorophenyl-2-propylamine,2-fa,o-fluoro-a-methylphenethylamine,benzeneethanamine,2-fluoro-a-methyl,1-2-fluorophenyl propan-2-aminehydrochloride PubChem CID: 121531 IUPAC Name: 1-(2-fluorophenyl)propan-2-amine SMILES: CC(N)CC1=CC=CC=C1F