missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorophenyl)-2-propylamine, 97%

Catalog No. AAH50999MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50999MD 250 mg
AAH5099903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50999MD Supplier Thermo Scientific Chemicals Supplier No. H50999MD

CAS: 1716-60-5 | C9H12FN | 153.20 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly Soluble (2.0 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1716-60-5
Molecular Formula C9H12FN
Molecular Weight (g/mol) 153.20
MDL Number MFCD01708100
InChI Key GDSXNLDTQFFIEU-UHFFFAOYNA-N
Synonym 2-fluoroamphetamine, 1-2-fluorophenyl propan-2-amine, o-fluoro-alpha-methylphenethylamine, phenethylamine, o-fluoro-alpha-methyl, 1-2-fluorophenyl-2-propylamine, 2-fa, o-fluoro-a-methylphenethylamine, benzeneethanamine,2-fluoro-a-methyl, 1-2-fluorophenyl propan-2-aminehydrochloride
PubChem CID 121531
IUPAC Name 1-(2-fluorophenyl)propan-2-amine
SMILES CC(N)CC1=CC=CC=C1F

Specifications

Quantity 250 mg
Solubility Information Slightly Soluble (2.0g/L) (25°C).
Formula Weight 153.2
Percent Purity 97%
Chemical Name or Material 1-(2-Fluorophenyl)-2-propylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber SH7030000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only