missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Epoxydodecane 95.0+%, TCI America™
Supplier: TCI America D1984100ML
Specifications
| 1,2-Epoxydodecane | |
| Colorless | |
| C12H24O | |
| 100 mL | |
| MPGABYXKKCLIRW-UHFFFAOYSA-N | |
| 2-decyloxirane | |
| 17858 | |
| ≥95.0% (GC) |
| 2855-19-8 | |
| 85°C | |
| MFCD00005150 | |
| 1,2-epoxydodecane, 1-dodecene oxide, oxirane, decyl, decyloxirane, decyl oxirane, dodecene epoxide, 1,2-dodecane oxide, 1,2-dodecene oxide, 1,2-epoxy-n-dodecene, nedox 1200 | |
| CCCCCCCCCCC1CO1 | |
| 184.323 | |
| 184.32 | |
| Liquid |
Chemical Identifiers
| 2855-19-8 | |
| 184.323 | |
| MPGABYXKKCLIRW-UHFFFAOYSA-N | |
| 17858 | |
| CCCCCCCCCCC1CO1 |
| C12H24O | |
| MFCD00005150 | |
| 1,2-epoxydodecane, 1-dodecene oxide, oxirane, decyl, decyloxirane, decyl oxirane, dodecene epoxide, 1,2-dodecane oxide, 1,2-dodecene oxide, 1,2-epoxy-n-dodecene, nedox 1200 | |
| 2-decyloxirane |
Safety and Handling
EINECSNumber : (2)-0220
RTECSNumber : JR2450000
TSCA : Yes