missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxydodecane 95.0+%, TCI America™

Catalog No. p-7127050
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D198425ML 25 mL
D1984100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D198425ML Supplier TCI America Supplier No. D198425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2855-19-8
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.323
MDL Number MFCD00005150
InChI Key MPGABYXKKCLIRW-UHFFFAOYSA-N
Synonym 1,2-epoxydodecane, 1-dodecene oxide, oxirane, decyl, decyloxirane, decyl oxirane, dodecene epoxide, 1,2-dodecane oxide, 1,2-dodecene oxide, 1,2-epoxy-n-dodecene, nedox 1200
PubChem CID 17858
IUPAC Name 2-decyloxirane
SMILES CCCCCCCCCCC1CO1

Specifications

Color Colorless
Boiling Point 85°C
Quantity 25 mL
Formula Weight 184.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxydodecane
EINECSNumber (2)-0220
RTECSNumber JR2450000
TSCA Yes