accessibility menu, dialog, popup

UserName

H3CO

CAS RN 2855-19-8

IUPAC Name: 2-decyloxirane
Synonyms: 2-decyloxirane
Molecular Weight (g/mol): 184.32
Molecular Formula: C12H24O
InChi Key: MPGABYXKKCLIRW-UHFFFAOYNA-N
SMILES: CCCCCCCCCCC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results