Learn More
1,2-Dimethylnaphthalene, 98%
CAS: 573-98-8 | C12H12 | 156.228 g/mol
Supplier: Thermo Scientific Chemicals L0977403
Description
1,2-Dimethylnaphthalene is suitable reagent used to investigate the secondary organic aerosol (SOA) production via gas-phase photooxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Dimethylnaphthalene is suitable reagent used to investigate the secondary organic aerosol (SOA) production via gas-phase photooxidation.
Solubility
Difficult to mix in water.
Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.
Specifications
| 1,2-Dimethylnaphthalene | |
| 1.013 | |
| >110°C (230°F) | |
| C12H12 | |
| MFCD00004035 | |
| 2039376 | |
| Difficult to mix in water. | |
| CC1=C(C2=CC=CC=C2C=C1)C | |
| 156.228 | |
| CHEBI:34052 | |
| 98% |
| 573-98-8 | |
| 266°C to 267°C | |
| Characteristic | |
| 1.616 | |
| 1 g | |
| dimethylnaphthalene, naphthalene, dimethyl, naphthalene, 1,2-dimethyl, 1,2-dimethyl naphthalene, unii-23t7o135bd, 1,2-dmn, 1,2-dimethyl-naphthalene, dsstox_cid_5130, dsstox_rid_77675, dsstox_gsid_25130 | |
| QNLZIZAQLLYXTC-UHFFFAOYSA-N | |
| 1,2-dimethylnaphthalene | |
| 11317 | |
| 156.23 |
Chemical Identifiers
| 573-98-8 | |
| 156.228 | |
| QNLZIZAQLLYXTC-UHFFFAOYSA-N | |
| 11317 | |
| 1,2-dimethylnaphthalene |
| C12H12 | |
| MFCD00004035 | |
| dimethylnaphthalene, naphthalene, dimethyl, naphthalene, 1,2-dimethyl, 1,2-dimethyl naphthalene, unii-23t7o135bd, 1,2-dmn, 1,2-dimethyl-naphthalene, dsstox_cid_5130, dsstox_rid_77675, dsstox_gsid_25130 | |
| CHEBI:34052 | |
| CC1=C(C2=CC=CC=C2C=C1)C |
Safety and Handling
EINECSNumber : 209-364-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only