accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 573-98-8

IUPAC Name: 1,2-dimethylnaphthalene
Synonyms: 1,2-dimethylnaphthalene dimethylnaphthalene (mixture of isomers)
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: QNLZIZAQLLYXTC-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=CC2=C1C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)