missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethylnaphthalene, 98%

Catalog No. p-7045197
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL09774MD 250 mg
AAL0977403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL09774MD Supplier Thermo Scientific Chemicals Supplier No. L09774MD

CAS: 573-98-8 | C12H12 | 156.228 g/mol

1,2-Dimethylnaphthalene is suitable reagent used to investigate the secondary organic aerosol (SOA) production via gas-phase photooxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dimethylnaphthalene is suitable reagent used to investigate the secondary organic aerosol (SOA) production via gas-phase photooxidation.

Solubility
Difficult to mix in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.

Chemical Identifiers

CAS 573-98-8
Molecular Formula C12H12
Molecular Weight (g/mol) 156.228
MDL Number MFCD00004035
InChI Key QNLZIZAQLLYXTC-UHFFFAOYSA-N
Synonym dimethylnaphthalene, naphthalene, dimethyl, naphthalene, 1,2-dimethyl, 1,2-dimethyl naphthalene, unii-23t7o135bd, 1,2-dmn, 1,2-dimethyl-naphthalene, dsstox_cid_5130, dsstox_rid_77675, dsstox_gsid_25130
PubChem CID 11317
ChEBI CHEBI:34052
IUPAC Name 1,2-dimethylnaphthalene
SMILES CC1=C(C2=CC=CC=C2C=C1)C

Specifications

Density 1.013
Boiling Point 266°C to 267°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.616
Quantity 250 mg
Beilstein 2039376
Solubility Information Difficult to mix in water.
Formula Weight 156.23
Percent Purity 98%
Chemical Name or Material 1,2-Dimethylnaphthalene
Show More Show Less
EINECSNumber 209-364-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only