missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimethylindole 98.0+%, TCI America™
Supplier: TCI America D139125G
Specifications
| 1,2-Dimethylindole | |
| 55°C | |
| C10H11N | |
| 25 g | |
| BJMUOUXGBFNLSN-UHFFFAOYSA-N | |
| 1,2-dimethylindole | |
| 13408 | |
| ≥98.0% (GC) |
| 875-79-6 | |
| 143°C | |
| MFCD00005802 | |
| 1,2-dimethyl-1h-indole, n-methyl-2-methylindole, indole, 1,2-dimethyl, 1h-indole, 1,2-dimethyl, unii-h373ts720o, dimethyl-1h-indole, 1,2-dimethylindolee, 1,2-dimethyl indole, pubchem7333, acmc-209qok | |
| CC1=CC2=CC=CC=C2N1C | |
| 145.205 | |
| 145.21 | |
| Crystalline Powder |
Chemical Identifiers
| 875-79-6 | |
| 145.205 | |
| BJMUOUXGBFNLSN-UHFFFAOYSA-N | |
| 13408 | |
| CC1=CC2=CC=CC=C2N1C |
| C10H11N | |
| MFCD00005802 | |
| 1,2-dimethyl-1h-indole, n-methyl-2-methylindole, indole, 1,2-dimethyl, 1h-indole, 1,2-dimethyl, unii-h373ts720o, dimethyl-1h-indole, 1,2-dimethylindolee, 1,2-dimethyl indole, pubchem7333, acmc-209qok | |
| 1,2-dimethylindole |
Safety and Handling
TSCA : No