missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethylindole 98.0+%, TCI America™

Catalog No. p-7126707
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13915G 5 g
D139125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13915G Supplier TCI America Supplier No. D13915G

Chemical Identifiers

CAS 875-79-6
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00005802
InChI Key BJMUOUXGBFNLSN-UHFFFAOYSA-N
Synonym 1,2-dimethyl-1h-indole, n-methyl-2-methylindole, indole, 1,2-dimethyl, 1h-indole, 1,2-dimethyl, unii-h373ts720o, dimethyl-1h-indole, 1,2-dimethylindolee, 1,2-dimethyl indole, pubchem7333, acmc-209qok
PubChem CID 13408
IUPAC Name 1,2-dimethylindole
SMILES CC1=CC2=CC=CC=C2N1C

Specifications

Melting Point 55°C
Boiling Point 143°C
Quantity 5 g
Formula Weight 145.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dimethylindole