missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluoro-4-iodobenzene (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. D256010G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D256010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D256010G Supplier TCI America Supplier No. D256010G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64248-58-4
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.99
MDL Number MFCD00044630
InChI Key KSASJELKLBIMSG-UHFFFAOYSA-N
Synonym 3,4-difluoroiodobenzene, 1,2-difluoro-4-iodo-benzene, benzene, 1,2-difluoro-4-iodo, 3,4-difluoro iodobenzene, pubchem3461, 3,4-difluoro-iodobenzene, 3,4-difluorophenyl iodide, 3,4-difluoro-1-iodobenzene, 1-iodo-3,4-difluoro-benzene
PubChem CID 2724444
IUPAC Name 1,2-difluoro-4-iodobenzene
SMILES FC1=C(F)C=C(I)C=C1

Specifications

Color Yellow
Quantity 10 g
Formula Weight 239.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Difluoro-4-iodobenzene (stabilized with Copper chip)