accessibility menu, dialog, popup

UserName

CAS RN 64248-58-4

IUPAC Name: 1,2-difluoro-4-iodobenzene
Synonyms: 1,2-difluoro-4-iodobenzene
Molecular Weight (g/mol): 239.99
Molecular Formula: C6H3F2I
InChi Key: KSASJELKLBIMSG-UHFFFAOYSA-N
SMILES: FC1=C(F)C=C(I)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)