missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluoro-4-iodobenzene, 99%

Catalog No. p-7081388
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1133906 5 g
AAA1133914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1133906 Supplier Thermo Scientific Chemicals Supplier No. A1133906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64248-58-4 | C6H3F2I | 239.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64248-58-4
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.99
MDL Number MFCD00044630
InChI Key KSASJELKLBIMSG-UHFFFAOYSA-N
Synonym 3,4-difluoroiodobenzene, 1,2-difluoro-4-iodo-benzene, benzene, 1,2-difluoro-4-iodo, 3,4-difluoro iodobenzene, pubchem3461, 3,4-difluoro-iodobenzene, 3,4-difluorophenyl iodide, 3,4-difluoro-1-iodobenzene, 1-iodo-3,4-difluoro-benzene
PubChem CID 2724444
IUPAC Name 1,2-difluoro-4-iodobenzene
SMILES FC1=C(F)C=C(I)C=C1

Specifications

Density 1.99
Boiling Point 177°C to 178°C
Flash Point 72°C (161°F)
Refractive Index 1.559
Quantity 5 g
Beilstein 2081099
Sensitivity Light sensitive
Formula Weight 239.99
Percent Purity 99%
Chemical Name or Material 1,2-Difluoro-4-iodobenzene
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only