missing translation for 'onlineSavingsMsg'
Learn More

1,2-Difluoro-4,5-dimethoxybenzene 95.0+%, TCI America™

Catalog No. D36265G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36265G 5 g
D36261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36265G Supplier TCI America Supplier No. D36265G
Only null left

Chemical Identifiers

CAS 203059-80-7
Molecular Formula C8H8F2O2
Molecular Weight (g/mol) 174.147
MDL Number MFCD00070789
InChI Key WWAOVLXLTJXDGS-UHFFFAOYSA-N
Synonym 4,5-difluoroveratrole, 4,5-difluoroveratrol, pubchem3039, acmc-1ccgk, 1,2-difluoro-4,5-dimethoxy-benzene, benzene,1,2-difluoro-4,5-dimethoxy, 1,2-bis fluoranyl-4,5-dimethoxy-benzene, 1,2-difluoro-4,5-dimethoxybenzene
PubChem CID 853175
IUPAC Name 1,2-difluoro-4,5-dimethoxybenzene
SMILES COC1=CC(=C(C=C1OC)F)F

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.15
Percent Purity ≥95.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,2-Difluoro-4,5-dimethoxybenzene