accessibility menu, dialog, popup

UserName

H3COFFOCH3

CAS RN 203059-80-7

IUPAC Name: 1,2-difluoro-4,5-dimethoxybenzene
Synonyms: 1,2-difluoro-4,5-dimethoxybenzene
Molecular Weight (g/mol): 174.15
Molecular Formula: C8H8F2O2
InChi Key: WWAOVLXLTJXDGS-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(F)C=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results