missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diethoxybenzene 98.0+%, TCI America™
Supplier: TCI America D045225G
Specifications
| 1,2-Diethoxybenzene | |
| 44°C | |
| 220°C | |
| MFCD00015143 | |
| o-diethoxybenzene, benzene, 1,2-diethoxy, catechol diethyl ether, benzene, o-diethoxy, o-diethoxy benzene, benzene, diethoxy, unii-p9dm5t78pz, pyrocatechol diethyl ether, p9dm5t78pz, diethoxybenzene-1,2 | |
| CCOC1=CC=CC=C1OCC | |
| 166.22 | |
| 166.22 | |
| Crystalline Powder |
| 2050-46-6 | |
| White | |
| C10H14O2 | |
| 25 g | |
| QZYDOKBVZJLQCK-UHFFFAOYSA-N | |
| 1,2-diethoxybenzene | |
| 74904 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2050-46-6 | |
| 166.22 | |
| QZYDOKBVZJLQCK-UHFFFAOYSA-N | |
| 74904 | |
| CCOC1=CC=CC=C1OCC |
| C10H14O2 | |
| MFCD00015143 | |
| o-diethoxybenzene, benzene, 1,2-diethoxy, catechol diethyl ether, benzene, o-diethoxy, o-diethoxy benzene, benzene, diethoxy, unii-p9dm5t78pz, pyrocatechol diethyl ether, p9dm5t78pz, diethoxybenzene-1,2 | |
| 1,2-diethoxybenzene |
Safety and Handling
RTECSNumber : CZ5580000
TSCA : Yes