accessibility menu, dialog, popup

UserName

CAS RN 2050-46-6

CH3OOH3C

CAS RN 2050-46-6

IUPAC Name: 1,2-diethoxybenzene
Synonyms: benzene, 1,2-diethoxy-
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: QZYDOKBVZJLQCK-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results