missing translation for 'onlineSavingsMsg'
Learn More

1,2-dichlorotetrafluoroethane (Freon 114), Restek™

Catalog No. 06728896
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06728896 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06728896 Supplier Chromatographic Supplier No. RK 30476
1mL/ampul contains 2000μg/mL in P and T Methanol
  • Methanol (99.8%)
  • 1,2- dichlorotetrafluoroethane(0.2%)

Chemical Identifiers

CAS 76-14-2 , 67-56-1
Molecular Formula C2Cl2F4
Molecular Weight (g/mol) 170.916
InChI Key DDMOUSALMHHKOS-UHFFFAOYSA-N
PubChem CID 6429
IUPAC Name 1,2-dichloro-1,1,2,2-tetrafluoroethane
SMILES C(C(F)(F)Cl)(F)(F)Cl

Specifications

Packaging Ampule
Explosion Limit Upper Limit: 36% Lower limit: 6%
Ignition Point 464°C
Odor Mild
Vapor Density 1.1 (air = 1)
Flash Point 52°C
Specific Gravity 0.791 to 0.792g/cm3 at 20°C
Physical Form Liquid
Concentration 2000 μg/mL in P and T Methanol
UN Number 1230
Melting Point -98°C
Solubility Moderately soluble
Type Single-component Standard
Quantity 1 mL
Formula Weight 32.04
Show More Show Less
Hazard Category Danger
Hazard Statement Highly flammable liquid and vapour.
Toxic if swallowed, in contact with skin or if inhaled.
Toxic if inhaled. Causes damage to organs.
Harms public health and the environment by destroying ozone in the upper atmosphere.
Precautionary Statement IF SWALLOWED: Immediately call a POISON CENTER/doctor/.
IF ON SKIN: Wash with plenty of soap and water.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed: Call a POISON CENTER or doctor/physician
ShelfLife 6 months
DOTInformation Class 3 (6.1): Packing Group II
EINECSNumber 200-659-6 (Methanol)