CAS RN 76-14-2
IUPAC Name:
1,2-dichloro-1,1,2,2-tetrafluoroethane
Synonyms:
1,2-dichlorotetrafluoroethane
Molecular Weight (g/mol):
170.92
Molecular Formula:
C2Cl2F4
InChi Key:
DDMOUSALMHHKOS-UHFFFAOYSA-N
SMILES:
FC(F)(Cl)C(F)(F)Cl
1
–
1
of
1
results
1
1,2-dichlorotetrafluoroethane (Freon 114), Restek™
CAS: 76-14-2 Molecular Formula: C2Cl2F4 Molecular Weight (g/mol): 170.916 InChI Key: DDMOUSALMHHKOS-UHFFFAOYSA-N PubChem CID: 6429 IUPAC Name: 1,2-dichloro-1,1,2,2-tetrafluoroethane SMILES: C(C(F)(F)Cl)(F)(F)Cl