missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-fluoro-5-nitrobenzene 98.0+%, TCI America™

Catalog No. p-7128410
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D46945G 5 g
D469425G 25 g
2 options

Catalog No. D46945G

Supplier: TCI America D46945G

Only null left

Chemical Identifiers

CAS 2339-78-8
Molecular Formula C6H2Cl2FNO2
Molecular Weight (g/mol) 209.985
MDL Number MFCD00075330
InChI Key FXOCDIKCKFOUDE-UHFFFAOYSA-N
Synonym 4,5-dichloro-2-fluoronitrobenzene, benzene, 1,2-dichloro-4-fluoro-5-nitro, buttpark 89\07-58, 4,5-dichloro-2-fluoro-1-nitrobenzene, pubchem4385, intermediates-zcf02626, ksc201k5p, 1,2-ichloro-4-luoro-5-itro-benzene
PubChem CID 533951
IUPAC Name 1,2-dichloro-4-fluoro-5-nitrobenzene
SMILES C1=C(C(=CC(=C1Cl)Cl)F)[N+](=O)[O-]

Specifications

Melting Point 17°C
Boiling Point 247°C
Quantity 5 g
Formula Weight 209.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dichloro-4-fluoro-5-nitrobenzene