accessibility menu, dialog, popup

UserName

ClClFNOO

CAS RN 2339-78-8

IUPAC Name: 1,2-dichloro-4-fluoro-5-nitrobenzene
Synonyms: 1,2-dichloro-4-fluoro-5-nitrobenzene
Molecular Weight (g/mol): 209.99
Molecular Formula: C6H2Cl2FNO2
InChi Key: FXOCDIKCKFOUDE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(Cl)C=C1F
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)