missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-fluoro-5-nitrobenzene, 95%

Catalog No. p-7056042
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6478206 5 g
AAH6478214 25 g
AAH6478222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6478206 Supplier Thermo Scientific Chemicals Supplier No. H6478206
Only null left

CAS: 2339-78-8 | C6H2Cl2FNO2 | 209.985 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2339-78-8
Molecular Formula C6H2Cl2FNO2
Molecular Weight (g/mol) 209.985
MDL Number MFCD00075330
InChI Key FXOCDIKCKFOUDE-UHFFFAOYSA-N
Synonym 4,5-dichloro-2-fluoronitrobenzene, benzene, 1,2-dichloro-4-fluoro-5-nitro, buttpark 89\07-58, 4,5-dichloro-2-fluoro-1-nitrobenzene, pubchem4385, intermediates-zcf02626, ksc201k5p, 1,2-ichloro-4-luoro-5-itro-benzene
PubChem CID 533951
IUPAC Name 1,2-dichloro-4-fluoro-5-nitrobenzene
SMILES C1=C(C(=CC(=C1Cl)Cl)F)[N+](=O)[O-]

Specifications

Melting Point 17°C
Color Yellow
Density 1.596 g/mL
Boiling Point 247°C
Flash Point >113°C (235°F)
Refractive Index 1.575
Quantity 5 g
Beilstein 3269124
Formula Weight 209.99
Percent Purity 95%
Physical Form Low Melting Solid
Chemical Name or Material 1,2-Dichloro-4-fluoro-5-nitrobenzene
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only