missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-5-chloro-3-fluorobenzene, 98%

CAS: 208186-78-1 | C6H2Br2ClF | 288.34 g/mol

$111.04 - $404.26

Chemical Identifiers

CAS 208186-78-1
Molecular Formula C6H2Br2ClF
Molecular Weight (g/mol) 288.34
MDL Number MFCD00143441
InChI Key XLDRDGJJGGYJCO-UHFFFAOYSA-N
Synonym 5-chloro-2,3-dibromo-1-fluorobenzene, 1-chloro-3,4-dibromo-5-fluorobenzene, 5-chloro-2,3-dibromofluorobenzene, 5-chloro-1,2-dibromo-3-fluorobenzene, 1,2-dibromo-5-chloro-3-fluoro-benzene, acmc-1cpco, benzene,1,2-dibromo-5-chloro-3-fluoro, 3,4-dibromo-5-fluorochlorobenzene, 3-fluoro-5-chloro-1,2-phenylene dibromide, 1,2-bis bromanyl-5-chloranyl-3-fluoranyl-benzene
PubChem CID 2724907
IUPAC Name 1,2-dibromo-5-chloro-3-fluorobenzene
SMILES FC1=CC(Cl)=CC(Br)=C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1953606
View Documents
Thermo Scientific Chemicals
A1953606
5 g
Each for $111.04
Only null left
 
AAA1953614
View Documents
Thermo Scientific Chemicals
A1953614
25 g
Each for $404.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

208186-78-1
288.34
XLDRDGJJGGYJCO-UHFFFAOYSA-N
2724907
FC1=CC(Cl)=CC(Br)=C1Br
C6H2Br2ClF
MFCD00143441
5-chloro-2,3-dibromo-1-fluorobenzene, 1-chloro-3,4-dibromo-5-fluorobenzene, 5-chloro-2,3-dibromofluorobenzene, 5-chloro-1,2-dibromo-3-fluorobenzene, 1,2-dibromo-5-chloro-3-fluoro-benzene, acmc-1cpco, benzene,1,2-dibromo-5-chloro-3-fluoro, 3,4-dibromo-5-fluorochlorobenzene, 3-fluoro-5-chloro-1,2-phenylene dibromide, 1,2-bis bromanyl-5-chloranyl-3-fluoranyl-benzene
1,2-dibromo-5-chloro-3-fluorobenzene

Specifications

208186-78-1
C6H2Br2ClF
5 g
XLDRDGJJGGYJCO-UHFFFAOYSA-N
1,2-dibromo-5-chloro-3-fluorobenzene
2724907
98%
37°C to 39°C
MFCD00143441
5-chloro-2,3-dibromo-1-fluorobenzene, 1-chloro-3,4-dibromo-5-fluorobenzene, 5-chloro-2,3-dibromofluorobenzene, 5-chloro-1,2-dibromo-3-fluorobenzene, 1,2-dibromo-5-chloro-3-fluoro-benzene, acmc-1cpco, benzene,1,2-dibromo-5-chloro-3-fluoro, 3,4-dibromo-5-fluorochlorobenzene, 3-fluoro-5-chloro-1,2-phenylene dibromide, 1,2-bis bromanyl-5-chloranyl-3-fluoranyl-benzene
FC1=CC(Cl)=CC(Br)=C1Br
288.34
288.35
1,2-Dibromo-5-chloro-3-fluorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only