accessibility menu, dialog, popup

UserName

BrBrFCl

CAS RN 208186-78-1

IUPAC Name: 1,2-dibromo-5-chloro-3-fluorobenzene
Synonyms: 1,2-dibromo-5-chloro-3-fluorobenzene
Molecular Weight (g/mol): 288.34
Molecular Formula: C6H2Br2ClF
InChi Key: XLDRDGJJGGYJCO-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(Br)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results