missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-5-chloro-3-fluorobenzene, 98%

Catalog No. AAA1953614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1953614 25 g
AAA1953606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1953614 Supplier Thermo Scientific Chemicals Supplier No. A1953614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 208186-78-1 | C6H2Br2ClF | 288.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 208186-78-1
Molecular Formula C6H2Br2ClF
Molecular Weight (g/mol) 288.34
MDL Number MFCD00143441
InChI Key XLDRDGJJGGYJCO-UHFFFAOYSA-N
Synonym 5-chloro-2,3-dibromo-1-fluorobenzene, 1-chloro-3,4-dibromo-5-fluorobenzene, 5-chloro-2,3-dibromofluorobenzene, 5-chloro-1,2-dibromo-3-fluorobenzene, 1,2-dibromo-5-chloro-3-fluoro-benzene, acmc-1cpco, benzene,1,2-dibromo-5-chloro-3-fluoro, 3,4-dibromo-5-fluorochlorobenzene, 3-fluoro-5-chloro-1,2-phenylene dibromide, 1,2-bis bromanyl-5-chloranyl-3-fluoranyl-benzene
PubChem CID 2724907
IUPAC Name 1,2-dibromo-5-chloro-3-fluorobenzene
SMILES FC1=CC(Cl)=CC(Br)=C1Br

Specifications

Melting Point 37°C to 39°C
Quantity 25 g
Formula Weight 288.35
Percent Purity 98%
Chemical Name or Material 1,2-Dibromo-5-chloro-3-fluorobenzene
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only