missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-4-fluorobenzene 98.0+%, TCI America™

Supplier:  TCI America D28545G

 View more versions of this product

Catalog No. D28545G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,2-Dibromo-4-fluorobenzene
Yellow
MFCD00070780
3,4-dibromofluorobenzene, 3,4-dibromo-1-fluorobenzene, pubchem3445, acmc-1ccqu, intermediates-zcf02634, 1,2-dibromo-4-fluorobezene, benzene,1,2-dibromo-4-fluoro, 1,2-dibromo-4-fluorobenzene, benzene, 1,2-dibromo-4-fluoro
C1=CC(=C(C=C1F)Br)Br
253.896
253.90
Liquid
2369-37-1
C6H3Br2F
5 g
RNTGKISRXVFIIP-UHFFFAOYSA-N
1,2-dibromo-4-fluorobenzene
75402
≥98.0% (GC)

Chemical Identifiers

2369-37-1
253.896
RNTGKISRXVFIIP-UHFFFAOYSA-N
75402
C1=CC(=C(C=C1F)Br)Br
C6H3Br2F
MFCD00070780
3,4-dibromofluorobenzene, 3,4-dibromo-1-fluorobenzene, pubchem3445, acmc-1ccqu, intermediates-zcf02634, 1,2-dibromo-4-fluorobezene, benzene,1,2-dibromo-4-fluoro, 1,2-dibromo-4-fluorobenzene, benzene, 1,2-dibromo-4-fluoro
1,2-dibromo-4-fluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions