missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-4-fluorobenzene 98.0+%, TCI America™

Catalog No. p-7127541
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28545G 5 g
D285425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28545G Supplier TCI America Supplier No. D28545G
Only null left

Chemical Identifiers

CAS 2369-37-1
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.896
MDL Number MFCD00070780
InChI Key RNTGKISRXVFIIP-UHFFFAOYSA-N
Synonym 3,4-dibromofluorobenzene, 3,4-dibromo-1-fluorobenzene, pubchem3445, acmc-1ccqu, intermediates-zcf02634, 1,2-dibromo-4-fluorobezene, benzene,1,2-dibromo-4-fluoro, 1,2-dibromo-4-fluorobenzene, benzene, 1,2-dibromo-4-fluoro
PubChem CID 75402
IUPAC Name 1,2-dibromo-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Br)Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 253.90
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dibromo-4-fluorobenzene