missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromophenyl)-1H-pyrazole, ≥97%, Thermo Scientific™

Catalog No. CC24410DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC24410DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC24410DA Supplier Thermo Scientific Chemicals Supplier No. CC24410DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87488-84-4
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.07
MDL Number MFCD06659065
InChI Key QNDJHGODPWAKAO-UHFFFAOYSA-N
Synonym 1-2-bromophenyl-1h-pyrazole, 1-2-bromophenyl pyrazole, 1-2-bromo-phenyl-1h-pyrazole, 2-bromophenyl pyrazole, acmc-209qo1, 1h-pyrazole,1-2-bromophenyl, 1h-pyrazole, 1-2-bromophenyl
PubChem CID 2795433
IUPAC Name 1-(2-bromophenyl)-1H-pyrazole
SMILES BrC1=CC=CC=C1N1C=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 1-(2-Bromophenyl)-1H-pyrazole
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.