missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one, 95%

Catalog No. AAH3652803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3652803 1 g
AAH3652806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3652803 Supplier Thermo Scientific Chemicals Supplier No. H3652803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84501-67-7 | C5H9BrN4O | 221.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84501-67-7
Molecular Formula C5H9BrN4O
Molecular Weight (g/mol) 221.058
MDL Number MFCD16251259
InChI Key KQKSCWCKLKLXJS-UHFFFAOYSA-N
Synonym 1-2-bromoethyl-4-ethyl-1,4-dihydro-5h-tetrazol-5-one, 1-2-bromoethyl-4-ethyltetrazol-5-one, 1-2-bromoethyl-4-ethyl-tetrazol-5-one, 1-2-bromoethyl-4-ethyl-1,2,3,4-tetrazol-5-one, 2-4-ethyl-4,5-dihydro-5-oxo-1h-tetrazol-1-yl ethyl bromide
PubChem CID 3019915
IUPAC Name 1-(2-bromoethyl)-4-ethyltetrazol-5-one
SMILES CCN1C(=O)N(N=N1)CCBr

Specifications

Color Colorless
Density 1.777 g/mL
Boiling Point 223°C
Flash Point 89°C (192°F)
Refractive Index 1.507
Quantity 1 g
Formula Weight 221.06
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-(2-Bromoethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one
Hazard Category H227-H302-H317-H318
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P370+P378q-P501c
EINECSNumber 282-986-4
TSCA No

RUO – Research Use Only