missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(4'-methyl-2,2'-bipyridin-4-yl)ethane 96.0+%, TCI America™

$190.59 - $569.85

Chemical Identifiers

CAS 96897-04-0
Molecular Formula C24H22N4
Molecular Weight (g/mol) 366.468
InChI Key KLTATORHZNMNDJ-UHFFFAOYSA-N
Synonym 4,4′C′C-(1,2-Ethanediyl)bis(4′C-methyl-2,2′C-bipyridine)
PubChem CID 15771932
IUPAC Name 2-(4-methylpyridin-2-yl)-4-[2-[2-(4-methylpyridin-2-yl)pyridin-4-yl]ethyl]pyridine
SMILES CC1=CC(=NC=C1)C2=NC=CC(=C2)CCC3=CC(=NC=C3)C4=NC=CC(=C4)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B3973200MG
View Documents
TCI America
B3973200MG
200 mg
Each for $190.59
Only null left
 
B39731G
View Documents
TCI America
B39731G
1 g
Each for $569.85
Only null left
 
Description

Description

Specifications

Chemical Identifiers

96897-04-0
366.468
4,4′C′C-(1,2-Ethanediyl)bis(4′C-methyl-2,2′C-bipyridine)
2-(4-methylpyridin-2-yl)-4-[2-[2-(4-methylpyridin-2-yl)pyridin-4-yl]ethyl]pyridine
C24H22N4
KLTATORHZNMNDJ-UHFFFAOYSA-N
15771932
CC1=CC(=NC=C1)C2=NC=CC(=C2)CCC3=CC(=NC=C3)C4=NC=CC(=C4)C

Specifications

96897-04-0
Yellow
200 mg
KLTATORHZNMNDJ-UHFFFAOYSA-N
2-(4-methylpyridin-2-yl)-4-[2-[2-(4-methylpyridin-2-yl)pyridin-4-yl]ethyl]pyridine
15771932
≥96.0% (GC)
1,2-Bis(4′-methyl-2,2′-bipyridin-4-yl)ethane
192°C
C24H22N4
4,4′C′C-(1,2-Ethanediyl)bis(4′C-methyl-2,2′C-bipyridine)
CC1=CC(=NC=C1)C2=NC=CC(=C2)CCC3=CC(=NC=C3)C4=NC=CC(=C4)C
366.468
366.47
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents