missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(4'-methyl-2,2'-bipyridin-4-yl)ethane 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 192°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 366.47 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2-Bis(4′-methyl-2,2′-bipyridin-4-yl)ethane |
Safety and Handling
| TSCA | No |