missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(4'-methyl-2,2'-bipyridin-4-yl)ethane 96.0+%, TCI America™

Catalog No. b3973200mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
B3973200MG 200 mg
B39731G 1 g
2 options

Catalog No. B3973200MG

Supplier: TCI America B3973200MG

Only null left

Chemical Identifiers

CAS 96897-04-0
Molecular Formula C24H22N4
Molecular Weight (g/mol) 366.468
InChI Key KLTATORHZNMNDJ-UHFFFAOYSA-N
Synonym 4,4′C′C-(1,2-Ethanediyl)bis(4′C-methyl-2,2′C-bipyridine)
PubChem CID 15771932
IUPAC Name 2-(4-methylpyridin-2-yl)-4-[2-[2-(4-methylpyridin-2-yl)pyridin-4-yl]ethyl]pyridine
SMILES CC1=CC(=NC=C1)C2=NC=CC(=C2)CCC3=CC(=NC=C3)C4=NC=CC(=C4)C

Specifications

Melting Point 192°C
Color Yellow
Quantity 200 mg
Formula Weight 366.47
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(4′-methyl-2,2′-bipyridin-4-yl)ethane