missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(3,4-dimethylphenyl)ethane 95.0+%, TCI America™

Catalog No. p-7119304
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18511G 1 g
B18515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18511G Supplier TCI America Supplier No. B18511G
Only null left

Chemical Identifiers

CAS 34101-86-5
Molecular Formula C18H22
Molecular Weight (g/mol) 238.37
MDL Number MFCD00037055
InChI Key MOPBWASVAUDDTC-UHFFFAOYSA-N
Synonym 1,2-Di(3,4-xylyl)ethane
PubChem CID 36676
IUPAC Name 4-[2-(3,4-dimethylphenyl)ethyl]-1,2-dimethylbenzene
SMILES CC1=CC=C(CCC2=CC=C(C)C(C)=C2)C=C1C

Specifications

Melting Point 94°C
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 238.37
Percent Purity ≥95.0% (GC)
Chemical Name or Material 1,2-Bis(3,4-dimethylphenyl)ethane
EINECSNumber (4)-1667
TSCA Yes