CAS RN 34101-86-5
CAS RN 34101-86-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2-Bis(3,4-dimethylphenyl)ethane 95.0+%, TCI America™
CAS: 34101-86-5 Molecular Formula: C18H22 Molecular Weight (g/mol): 238.37 MDL Number: MFCD00037055 InChI Key: MOPBWASVAUDDTC-UHFFFAOYSA-N Synonym: 1,2-Di(3,4-xylyl)ethane PubChem CID: 36676 IUPAC Name: 4-[2-(3,4-dimethylphenyl)ethyl]-1,2-dimethylbenzene SMILES: CC1=CC=C(CCC2=CC=C(C)C(C)=C2)C=C1C