missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzofluorene 95.0+%, TCI America™
$346.71 - $1187.40
Chemical Identifiers
| CAS | 238-84-6 |
|---|---|
| Molecular Formula | C17H12 |
| Molecular Weight (g/mol) | 216.283 |
| MDL Number | MFCD00039578 |
| InChI Key | HKMTVMBEALTRRR-UHFFFAOYSA-N |
| Synonym | 1,2-benzofluorene, 11h-benzo a fluorene, chrysofluorene, benzo a fluorene, alpha-naphthofluorene, benzo a fluorene van, unii-65ep6zt62k, 2,3-benzopyrene |
| PubChem CID | 9195 |
| ChEBI | CHEBI:82402 |
| IUPAC Name | 11H-benzo[a]fluorene |
| SMILES | C1C2=CC=CC=C2C3=C1C4=CC=CC=C4C=C3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B3931200MG
|
TCI America
B3931200MG |
200 mg |
Each for $346.71
|
|
|||||
|
B39311G
|
TCI America
B39311G |
1 g |
Each for $1,187.40
|
|
|||||
Chemical Identifiers
| 238-84-6 | |
| 216.283 | |
| HKMTVMBEALTRRR-UHFFFAOYSA-N | |
| 9195 | |
| 11H-benzo[a]fluorene |
| C17H12 | |
| MFCD00039578 | |
| 1,2-benzofluorene, 11h-benzo a fluorene, chrysofluorene, benzo a fluorene, alpha-naphthofluorene, benzo a fluorene van, unii-65ep6zt62k, 2,3-benzopyrene | |
| CHEBI:82402 | |
| C1C2=CC=CC=C2C3=C1C4=CC=CC=C4C=C3 |
Specifications
| 238-84-6 | |
| 407°C | |
| MFCD00039578 | |
| 3077 | |
| HKMTVMBEALTRRR-UHFFFAOYSA-N | |
| 11H-benzo[a]fluorene | |
| 9195 | |
| 216.28 | |
| Crystalline Powder |
| 182°C | |
| C17H12 | |
| 200 mg | |
| 1,2-benzofluorene, 11h-benzo a fluorene, chrysofluorene, benzo a fluorene, alpha-naphthofluorene, benzo a fluorene van, unii-65ep6zt62k, 2,3-benzopyrene | |
| C1C2=CC=CC=C2C3=C1C4=CC=CC=C4C=C3 | |
| 216.283 | |
| CHEBI:82402 | |
| ≥95.0% (GC) | |
| 1,2-Benzofluorene |
Safety and Handling
RTECSNumber : DF6382000
TSCA : No