missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzofluorene 95.0+%, TCI America™

Catalog No. B3931200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3931200MG 200 mg
B39311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3931200MG Supplier TCI America Supplier No. B3931200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 238-84-6
Molecular Formula C17H12
Molecular Weight (g/mol) 216.283
MDL Number MFCD00039578
InChI Key HKMTVMBEALTRRR-UHFFFAOYSA-N
Synonym 1,2-benzofluorene, 11h-benzo a fluorene, chrysofluorene, benzo a fluorene, alpha-naphthofluorene, benzo a fluorene van, unii-65ep6zt62k, 2,3-benzopyrene
PubChem CID 9195
ChEBI CHEBI:82402
IUPAC Name 11H-benzo[a]fluorene
SMILES C1C2=CC=CC=C2C3=C1C4=CC=CC=C4C=C3

Specifications

Melting Point 182°C
Boiling Point 407°C
Quantity 200 mg
UN Number 3077
Formula Weight 216.28
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Benzofluorene
RTECSNumber DF6382000
TSCA No