Learn More
1,2-Benzenedithiol, 97%
CAS: 17534-15-5 | C6H6S2 | 142.23 g/mol
Supplier: Thermo Scientific Chemicals L1125303
Description
1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.
Solubility
Soluble in water, benzene, ethanol, diethyl ether, ethyl acetate.
Notes
Incompatible with bases and strong oxidizing agents.
Specifications
| 1,2-Benzenedithiol | |
| 21°C to 25°C | |
| 239°C | |
| Unpleasant | |
| 1.658 | |
| 1 g | |
| 636154 | |
| 1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiol | |
| JRNVQLOKVMWBFR-UHFFFAOYSA-N | |
| benzene-1,2-dithiol | |
| 69370 | |
| 97% |
| 17534-15-5 | |
| 1.236 | |
| 103°C (217°F) | |
| C6H6S2 | |
| MFCD00004835 | |
| UN3335 | |
| Air sensitive | |
| Soluble in water,benzene,ethanol,diethyl ether,ethyl acetate. | |
| SC1=C(S)C=CC=C1 | |
| 142.23 | |
| 142.24 |
Chemical Identifiers
| 17534-15-5 | |
| 142.23 | |
| JRNVQLOKVMWBFR-UHFFFAOYSA-N | |
| 69370 | |
| SC1=C(S)C=CC=C1 |
| C6H6S2 | |
| MFCD00004835 | |
| 1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiol | |
| benzene-1,2-dithiol |
Safety and Handling
GHS H Statement
H318-H302-H312-H332-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
EINECSNumber : 241-530-4
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only