missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedithiol, 97%

Catalog No. p-7045585
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11253MD 250 mg
AAL1125303 1 g
AAL1125306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL11253MD Supplier Thermo Scientific Chemicals Supplier No. L11253MD
Only null left

CAS: 17534-15-5 | C6H6S2 | 142.23 g/mol

1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Benzenedithiol is used in preparation of schiff base, which is employed to make new series of copper(II) and zinc(II) complexes.

Solubility
Soluble in water, benzene, ethanol, diethyl ether, ethyl acetate.

Notes
Incompatible with bases and strong oxidizing agents.

Chemical Identifiers

CAS 17534-15-5
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00004835
InChI Key JRNVQLOKVMWBFR-UHFFFAOYSA-N
Synonym 1,2-benzenedithiol, benzenedithiol, 1,2-dimercaptobenzene, dithiocatechol, dimercaptobenzene, o-benzenedithiol, acmc-209e9y, 1,2-benzenedithiol 9ci, benzene-1,2-dithiol
PubChem CID 69370
IUPAC Name benzene-1,2-dithiol
SMILES SC1=C(S)C=CC=C1

Specifications

Melting Point 21°C to 25°C
Density 1.236
Boiling Point 239°C
Flash Point 103°C (217°F)
Odor Unpleasant
Refractive Index 1.658
Quantity 250 mg
UN Number UN3335
Beilstein 636154
Sensitivity Air sensitive
Solubility Information Soluble in water,benzene,ethanol,diethyl ether,ethyl acetate.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material 1,2-Benzenedithiol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H318-H302-H312-H332-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
EINECSNumber 241-530-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only